4-(2-Amino-1,3-thiazol-4-yl)-2,6-dimethylbenzenol ≥95%

Supplier: Matrix Scientific

043971-500MG 043971-1G 043971-5G
101863-464EA 375.38 USD
101863-464 101863-466 101863-468
4-(2-Amino-1,3-thiazol-4-yl)-2,6-dimethylbenzenol ≥95%
4-(2-Amino-1,3-thiazol-4-yl)-2,6-dimethylbenzenol

Hazard: Irritant

Formula: C₁₁H₁₂N₂OS
Melting Pt: 126…130 °C
MDL Number: MFCD04117801
CAS Number: 863305-85-5

Order Now

Specification Test Results

Melting point 126-130°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR