Methyl-3-(2,4-dinitrophenoxy)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

043970-500MG 043970-1G 043970-5G
101863-458EA 375.38 USD
101863-458 101863-460 101863-462
Methyl-3-(2,4-dinitrophenoxy)-2-thiophenecarboxylate ≥95%
Methyl-3-(2,4-dinitrophenoxy)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₂H₈N₂O₇S
Melting Pt: 120…122 °C
MDL Number: MFCD04117800
CAS Number: 104636-76-2

Order Now

Specification Test Results

Melting point 120-122°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR