Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-2-(4-chlorophenyl)acetate hydrochloride

043969-500MG 043969-1G 043969-5G
101863-452EA 181.54 USD
101863-452 101863-454 101863-456
Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride ≥95%
Methyl-2-amino-2-(4-chlorophenyl)acetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₁Cl₂NO₂
MW: 236.1 g/mol
Melting Pt: 199…202 °C
MDL Number: MFCD00196831
CAS Number: 42718-19-4

Order Now

Specification Test Results

Melting point 199-202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR