2-{[(Benzyloxy)carbonyl]amino}-2-(4-chlorophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

043968-500MG 043968-1G 043968-5G
101863-446EA 400 USD
101863-446 101863-448 101863-450
2-{[(Benzyloxy)carbonyl]amino}-2-(4-chlorophenyl)acetic acid ≥95%
2-{[(Benzyloxy)carbonyl]amino}-2-(4-chlorophenyl)acetic acid

Hazard: Irritant

Formula: C₁₆H₁₄ClNO₄
Melting Pt: 130…133 °C
MDL Number: MFCD04117799
CAS Number: 69902-04-1

Order Now

Specification Test Results

Melting point 130-133°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR