1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine ≥95%

Supplier: Matrix Scientific

043965-500MG 043965-1G 043965-5G
101863-434EA 36.92 USD
101863-434 101863-436 101863-438
1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine ≥95%
1-Methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine

Hazard: Irritant

Formula: C₇H₈N₄
MW: 148.17 g/mol
Melting Pt: 104…106 °C
MDL Number: MFCD04117796
CAS Number: 72583-83-6

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR