Methyl-4'-chloro[1,1'-biphenyl]-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4'-chloro-[1,1'-biphenyl]-4-carboxylate

043960-500MG 043960-1G 043960-5G
101863-418EA 556.92 USD
101863-418 101863-420 101863-422
Methyl-4'-chloro[1,1'-biphenyl]-4-carboxylate ≥95%
Methyl-4'-chloro[1,1'-biphenyl]-4-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₁ClO₂
MW: 246.7 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD04117794
CAS Number: 89901-02-0

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR