Diethyl-2-{[(5-chloro-2-pyridinyl)amino]methylene}malonate ≥95%

Supplier: Matrix Scientific

043956-500MG 043956-1G 043956-5G
101863-396EA 181.54 USD
101863-396 101863-398 101863-400
Diethyl-2-{[(5-chloro-2-pyridinyl)amino]methylene}malonate ≥95%
Diethyl-2-{[(5-chloro-2-pyridinyl)amino]methylene}malonate

Hazard: Irritant

Formula: C₁₃H₁₅ClN₂O₄
Melting Pt: 135…136 °C
MDL Number: MFCD00487269
CAS Number: 16867-57-5

Order Now

Specification Test Results

Melting point 135-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR