6,8-Dibromo-2-methyl-3-quinolinol ≥95%

Supplier: Matrix Scientific

043954-500MG 043954-1G 043954-5G
101863-384EA 278.46 USD
101863-384 101863-386 101863-388
6,8-Dibromo-2-methyl-3-quinolinol ≥95%
6,8-Dibromo-2-methyl-3-quinolinol

Hazard: Irritant

Formula: C₁₀H₇Br₂NO
MW: 316.99 g/mol
Melting Pt: 252 °C (dec.)
MDL Number: MFCD03617848
CAS Number: 59869-01-1

Order Now

Specification Test Results

Melting point 252°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR