7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carbonitrile ≥95%
Supplier: Matrix Scientific
043953-500MG
043953-1G
101863-380EA
921.54
USD
101863-380
101863-382
7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carbonitrile ≥95%
7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carbonitrile
Hazard: Irritant
Formula:
C₁₅H₁₀F₃N₅ Melting Pt: 230…232 °C |
MDL Number:
MFCD03791200 CAS Number: 863305-83-3 |
Specification Test Results
Melting point | 230-232°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...