tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-(4-cyanophenyl)piperazine-1-carboxylate, 4-(Piperazin-4-yl)benzonitrile, N1-BOC protected
043950-500MG
043950-1G
043950-5G
101863-362EA
375.38
USD
101863-362
101863-364
101863-366
tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(4-cyanophenyl)tetrahydro-1(2H)-pyrazinecarboxylate
Hazard: Irritant
Formula:
C₁₆H₂₁N₃O₂ MW: 287.36 g/mol Melting Pt: 128…126 °C |
MDL Number:
MFCD03791198 CAS Number: 186650-98-6 |
Specification Test Results
Melting point | 128-126°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...