[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]methanol ≥95%

Supplier: Matrix Scientific

043945-500MG 043945-1G 043945-5G
101863-332EA 278.46 USD
101863-332 101863-334 101863-336
[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]methanol ≥95%
[2-(4-Chlorophenyl)-4-methyl-1,3-thiazol-5-yl]methanol

Hazard: Irritant

Formula: C₁₁H₁₀NOClS
Melting Pt: 146-148°C
MDL Number: MFCD03791195
CAS Number: 317319-22-5

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR