5-Morpholino-2-nitrobenzenecarboxamide ≥95%
Supplier: Matrix Scientific
043944-500MG
043944-1G
043944-5G
101863-326EA
375.38
USD
101863-326
101863-328
101863-330
5-Morpholino-2-nitrobenzenecarboxamide ≥95%
5-Morpholino-2-nitrobenzenecarboxamide
Hazard: Irritant
Formula:
C₁₁H₁₃N₃O₄ MW: 251.24 g/mol Melting Pt: 280…282 °C |
MDL Number:
MFCD03791194 CAS Number: 404009-38-7 |
Specification Test Results
Melting point | 280-282°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...