4-(2-Hydroxyethyl)-5-methyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one ≥95%
Supplier: Matrix Scientific
043943-500MG
043943-1G
043943-5G
101863-320EA
181.54
USD
101863-320
101863-322
101863-324
4-(2-Hydroxyethyl)-5-methyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one ≥95%
4-(2-Hydroxyethyl)-5-methyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₂ Melting Pt: 95…98 °C |
MDL Number:
MFCD03617960 CAS Number: 861210-10-8 |
Specification Test Results
Melting point | 95-98°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...