7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
043942-500MG
043942-1G
043942-5G
101863-314EA
420
USD
101863-314
101863-316
101863-318
7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylic acid ≥95%
7-Amino-2-methyl-3-[3-(trifluoromethyl)phenyl]pyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₁F₃N₄O₂ MW: 336.28 g/mol Melting Pt: 224…226 °C |
MDL Number:
MFCD03791193 CAS Number: 863305-82-2 |
Specification Test Results
Melting point | 224-226°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...