Ethyl-4-hydroxy-5-oxo-1-(2-thienylmethyl)-2,5-dihydro-1H-pyrrole-3-carboxylate ≥95%

Supplier: Matrix Scientific

043940-500MG 043940-1G 043940-5G
101863-302EA 181.54 USD
101863-302 101863-304 101863-306
Ethyl-4-hydroxy-5-oxo-1-(2-thienylmethyl)-2,5-dihydro-1H-pyrrole-3-carboxylate ≥95%
Ethyl-4-hydroxy-5-oxo-1-(2-thienylmethyl)-2,5-dihydro-1H-pyrrole-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₄S
MW: 267.31 g/mol
Melting Pt: 98…100 °C
MDL Number: MFCD03791192
CAS Number: 131436-78-7

Order Now

Specification Test Results

Melting point 98-100°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR