Ethyl-2-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
043934-500MG
043934-1G
043934-5G
101863-272EA
278.46
USD
101863-272
101863-274
101863-276
Ethyl-2-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylate ≥95%
Ethyl-2-[3-methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₀F₃N₃O₂S MW: 305.28 g/mol Melting Pt: 109…110 °C |
MDL Number:
MFCD03305618 CAS Number: 955975-47-0 |
Specification Test Results
Melting point | 109-110°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...