Ethyl-N-[(5-amino-1-methyl-1H-pyrazol-4-yl)carbonyl]ethanehydrazonoate ≥95%

Supplier: Matrix Scientific

043927-500MG 043927-1G
101863-248EA 278.46 USD
101863-248 101863-250
Ethyl-N-[(5-amino-1-methyl-1H-pyrazol-4-yl)carbonyl]ethanehydrazonoate ≥95%
Ethyl-N-[(5-amino-1-methyl-1H-pyrazol-4-yl)carbonyl]ethanehydrazonoate

Hazard: Irritant

Formula: C₉H₁₅N₅O₂
Melting Pt: 199…201 °C
MDL Number: MFCD05021852
CAS Number: 114936-20-8

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR