5-Nitro-2-(1-pyrrolidinyl)pyridine ≥95%

Supplier: Matrix Scientific

043925-500MG 043925-1G 043925-5G
101863-236EA 278.46 USD
101863-236 101863-238 101863-240
5-Nitro-2-(1-pyrrolidinyl)pyridine ≥95%
5-Nitro-2-(1-pyrrolidinyl)pyridine

Hazard: Irritant

Formula: C₉H₁₁N₃O₂
Melting Pt: 137…139 °C
MDL Number: MFCD00119573
CAS Number: 26820-63-3

Order Now

Specification Test Results

Melting point 137-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR