5-Amino-1-methyl-1H-pyrazole-4-carbohydrazide ≥95%
Supplier: Matrix Scientific
043924-500MG
043924-1G
043924-5G
101863-230EA
207.69
USD
101863-230
101863-232
101863-234
5-Amino-1-methyl-1H-pyrazole-4-carbohydrazide ≥95%
5-Amino-1-methyl-1H-pyrazole-4-carbohydrazide
Hazard: Irritant
Formula:
C₅H₉N₅O Melting Pt: 237…240 °C |
MDL Number:
MFCD03425741 CAS Number: 99347-21-4 |
Specification Test Results
Melting point | 237-240°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...