Ethyl-4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-methyl-2-(3-pyridyl)thiazole-5-carboxylate
043923-500MG
043923-1G
043923-5G
101863-224EA
375.38
USD
101863-224
101863-226
101863-228
Ethyl-4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate ≥95%
Ethyl 4-methyl-2-(3-pyridinyl)-1,3-thiazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₂S MW: 248.30 g/mol Melting Pt: 59-60°C |
MDL Number:
MFCD01567252 CAS Number: 39091-00-4 |
Specification Test Results
Melting point | 59-60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...