1-(3-Fluorobenzyl)-4-piperidinecarboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

043922-500MG 043922-1G 043922-5G
101863-218EA 375.38 USD
101863-218 101863-220 101863-222
1-(3-Fluorobenzyl)-4-piperidinecarboxylic acid hydrochloride ≥95%
1-(3-Fluorobenzyl)-4-piperidinecarboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₇ClFNO₂
MW: 273.74 g/mol
Melting Pt: 220…223 °C
MDL Number: MFCD03425740
CAS Number: 451485-55-5

Order Now

Specification Test Results

Melting point 220-223°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR