1-(3,4-Dichlorobenzyl)-4-piperidinecarboxylic acid hydrochloride ≥95%
Supplier: Matrix Scientific
043921-500MG
043921-1G
043921-5G
101863-212EA
375.38
USD
101863-212
101863-214
101863-216
1-(3,4-Dichlorobenzyl)-4-piperidinecarboxylic acid hydrochloride ≥95%
1-(3,4-Dichlorobenzyl)-4-piperidinecarboxylic acid hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₆Cl₃NO₂ Melting Pt: 260…262 °C |
MDL Number:
MFCD03425739 CAS Number: 451485-54-4 |
Specification Test Results
Melting point | 260-262°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...