2-[(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)carbonyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

043917-500MG 043917-1G 043917-5G
101863-190EA 278.46 USD
101863-190 101863-192 101863-194
2-[(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)carbonyl]benzenecarboxylic acid ≥95%
2-[(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)carbonyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₁NO₅
Melting Pt: 172…173 °C
MDL Number: MFCD03012223
CAS Number: 26513-80-4

Order Now

Specification Test Results

Melting point 172-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR