Ethyl-5-cyano-2,4-diphenyl-6-thioxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

043915-500MG 043915-1G 043915-5G
101863-184EA 278.46 USD
101863-184 101863-186 101863-188
Ethyl-5-cyano-2,4-diphenyl-6-thioxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate ≥95%
Ethyl-5-cyano-2,4-diphenyl-6-thioxo-1,4,5,6-tetrahydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₂₁H₁₈N₂O₂S
Melting Pt: 118…121 °C
MDL Number: MFCD03425736
CAS Number: 105199-50-6

Order Now

Specification Test Results

Melting point 118-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR