2-[3-(Trifluoromethyl)phenyl]ethanethioamide ≥97%

Supplier: Matrix Scientific

Synonyms: 2-[3-(Trifluoromethyl)phenyl]thioacetamide

043913-500MG 043913-1G
101863-174EA 161.54 USD
101863-174 101863-176
2-[3-(Trifluoromethyl)phenyl]ethanethioamide ≥97%
2-[3-(Trifluoromethyl)phenyl]ethanethioamide

Hazard: Irritant

Formula: C₉H₈F₃NS
MW: 219.23 g/mol
Melting Pt: 78…80 °C
MDL Number: MFCD09817423
CAS Number: 952182-78-4

Order Now

Specification Test Results

Melting point 78-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR