3-(1H-Pyrrol-1-yl)-1-benzothiophene-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

043910-500MG 043910-1G 043910-5G
101863-168EA 170.77 USD
101863-168 101863-170 101863-172
3-(1H-Pyrrol-1-yl)-1-benzothiophene-2-carbohydrazide ≥95%
3-(1H-Pyrrol-1-yl)-1-benzothiophene-2-carbohydrazide

Hazard: Irritant

Formula: C₁₃H₁₁N₃OS
Melting Pt: 129…132 °C
MDL Number: MFCD06200944
CAS Number: 107363-01-9

Order Now

Specification Test Results

Melting point 129-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR