2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1-Oxo-2,3-dihydro-1H-isoindol-2-yl)benzoic acid

043907-500MG 043907-1G 043907-5G
101863-156EA 375.38 USD
101863-156 101863-158 101863-160
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzenecarboxylic acid ≥95%
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₁NO₃
MW: 253.26 g/mol
Melting Pt: 240…243 °C
MDL Number: MFCD02186096
CAS Number: 4770-69-8

Order Now

Specification Test Results

Melting point 240-243°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR