Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-acetamido-2-hydroxybenzoate
043905-5G
043905-25G
043905-100G
101863-146EA
1000
USD
101863-146
101863-148
101863-150
Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate ≥95%
Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate
Hazard: Irritant
Formula:
C₁₀H₁₁NO₄ MW: 209.2 g/mol Melting Pt: 146…147 °C |
MDL Number:
MFCD00458326 CAS Number: 4093-28-1 |
Specification Test Results
Melting point | 146-147°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...