3-Bromo-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-7-azaindole

043903-500MG 043903-1G 043903-5G
101863-134EA 375.38 USD
101863-134 101863-136 101863-138
3-Bromo-1H-pyrrolo[2,3-b]pyridine ≥95%
3-Bromo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Melting Pt: 172…175 °C
MDL Number: MFCD02179276
CAS Number: 74420-15-8

Order Now

Specification Test Results

Melting point 172-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR