4-(4-Ethylanilino)-4-oxo-2-butenoic acid ≥95%

Supplier: Matrix Scientific

043901-500MG 043901-1G 043901-5G
101863-128EA 181.54 USD
101863-128 101863-130 101863-132
4-(4-Ethylanilino)-4-oxo-2-butenoic acid ≥95%
4-(4-Ethylanilino)-4-oxo-2-butenoic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
Melting Pt: 184…186 °C
MDL Number: MFCD00175103
CAS Number: 324067-34-7

Order Now

Specification Test Results

Melting point 184-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR