6-{[(2-Furylmethyl)amino]carbonyl}-3-cyclohexene-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

043900-500MG 043900-5G
101863-122EA 181.54 USD
101863-122 101863-126
6-{[(2-Furylmethyl)amino]carbonyl}-3-cyclohexene-1-carboxylic acid ≥95%
6-{[(2-Furylmethyl)amino]carbonyl}-3-cyclohexene-1-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
Melting Pt: 135…137 °C
MDL Number: MFCD00091304
CAS Number: 199443-38-4

Order Now

Specification Test Results

Melting point 135-137°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR