(1-Phenylethylidene)malononitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1-Phenylethylidene)propanedinitrile, 2-(1-Phenylethylidene)malononitrile

043899-500MG 043899-1G 043899-5G
101863-116EA 230.77 USD
101863-116 101863-118 101863-120
(1-Phenylethylidene)malononitrile ≥95%
(1-Phenylethylidene)malononitrile

Hazard: Irritant

Formula: C₁₁H₈N₂
MW: 168.1946 g/mol
Melting Pt: 94…96 °C
MDL Number: MFCD00159012
CAS Number: 5447-87-0

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR