2-Amino-1-butyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile ≥95%
Supplier: Matrix Scientific
043889-500MG
043889-1G
043889-5G
101863-080EA
278.46
USD
101863-080
101863-082
101863-084
2-Amino-1-butyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile ≥95%
2-Amino-1-butyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile
Hazard: Irritant
Formula:
C₂₁H₂₁N₃ Melting Pt: 124…126 °C |
MDL Number:
MFCD01815190 CAS Number: 477887-28-8 |
Specification Test Results
Melting point | 124-126°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...