{2-[4-(Benzyloxy)phenoxy]phenyl}methanol ≥95%

Supplier: Matrix Scientific

043888-500MG 043888-1G
101863-076EA 921.54 USD
101863-076 101863-078
{2-[4-(Benzyloxy)phenoxy]phenyl}methanol ≥95%
{2-[4-(Benzyloxy)phenoxy]phenyl}methanol

Hazard: Irritant

Formula: C₂₀H₁₈O₃
MW: 306.36 g/mol
Melting Pt: 75…77 °C
MDL Number: MFCD01814787
CAS Number: 449778-82-9

Order Now

Specification Test Results

Melting point 75-77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR