N-(1H-1,3-Benzimidazol-5-yl)thiourea ≥95%

Supplier: Matrix Scientific

043886-500MG 043886-1G 043886-5G
101863-066EA 375.38 USD
101863-066 101863-068 101863-070
N-(1H-1,3-Benzimidazol-5-yl)thiourea ≥95%
N-(1H-1,3-Benzimidazol-5-yl)thiourea

Hazard: Irritant

Formula: C₈H₈N₄S
Melting Pt: 214…216 °C
MDL Number: MFCD03791189
CAS Number: 862665-25-6

Order Now

Specification Test Results

Melting point 214-216°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR