2-(4-Chlorophenyl)-1-hydrazinecarbothioamide ≥95%
Supplier: Matrix Scientific
043884-500MG
043884-1G
043884-5G
101863-054EA
181.54
USD
101863-054
101863-056
101863-058
2-(4-Chlorophenyl)-1-hydrazinecarbothioamide ≥95%
2-(4-Chlorophenyl)-1-hydrazinecarbothioamide
Hazard: Irritant
Formula:
C₇H₈ClN₃S Melting Pt: 190…200 °C |
MDL Number:
MFCD00456275 CAS Number: 7382-41-4 |
Specification Test Results
Melting point | 190-200°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...