1,3-Benzoxazol-6-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Aminobenzoxazole, Benzo[d]oxazol-6-amine

043883-1G 043883-5G
101863-050EA 60 USD
101863-050 101863-052
1,3-Benzoxazol-6-amine ≥97%
1,3-Benzoxazol-6-amine

Hazard: Irritant/Store in Freezer

Formula: C₇H₆N₂O
MW: 134.10 g/mol
Melting Pt: 95…97 °C
MDL Number: MFCD03791187
CAS Number: 177492-52-3

Order Now

Specification Test Results

Melting point 95-97°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR