3-{2-[3-(Trifluoromethyl)phenoxy]phenyl}acrylic acid ≥95%

Supplier: Matrix Scientific

043879-500MG 043879-1G 043879-5G
101863-030EA 556.92 USD
101863-030 101863-032 101863-034
3-{2-[3-(Trifluoromethyl)phenoxy]phenyl}acrylic acid ≥95%
3-{2-[3-(Trifluoromethyl)phenoxy]phenyl}acrylic acid

Hazard: Irritant

Formula: C₁₆H₁₁F₃O₃
Melting Pt: 156…158 °C
MDL Number: MFCD01814780
CAS Number: 449778-81-8

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR