3-[2-(4-Iodophenoxy)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

043878-500MG 043878-1G
101863-026EA 375.38 USD
101863-026 101863-028
3-[2-(4-Iodophenoxy)phenyl]acrylic acid ≥95%
3-[2-(4-Iodophenoxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₅H₁₁IO₃
Melting Pt: 212…214 °C
MDL Number: MFCD02091016
CAS Number: 449778-80-7

Order Now

Specification Test Results

Melting point 212-214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR