2-(4-Fluorophenoxy)benzenecarbaldehyde oxime ≥95%
Supplier: Matrix Scientific
043875-500MG
043875-1G
043875-5G
101863-010EA
1109.23
USD
101863-010
101863-012
101863-014
2-(4-Fluorophenoxy)benzenecarbaldehyde oxime ≥95%
2-(4-Fluorophenoxy)benzenecarbaldehyde oxime
Hazard: Irritant
Formula:
C₁₃H₁₀FNO₂ Melting Pt: 107…109 °C |
MDL Number:
MFCD02091015 CAS Number: 449778-79-4 |
Specification Test Results
Melting point | 107-109°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...