2-(3-Chlorophenoxy)benzenecarbaldehyde oxime ≥95%

Supplier: Matrix Scientific

043873-500MG 043873-1G
101863-002EA 1109.23 USD
101863-002 101863-004
2-(3-Chlorophenoxy)benzenecarbaldehyde oxime ≥95%
2-(3-Chlorophenoxy)benzenecarbaldehyde oxime

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₂
Melting Pt: 85…87 °C
MDL Number: MFCD02091012
CAS Number: 449778-76-1

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR