4-Chloro-2-fluoro-5-(2-pyrimidinyloxy)aniline ≥95%

Supplier: Matrix Scientific

043872-500MG 043872-1G 043872-5G
101862-996EA 747.69 USD
101862-996 101862-998 101863-000
4-Chloro-2-fluoro-5-(2-pyrimidinyloxy)aniline ≥95%
4-Chloro-2-fluoro-5-(2-pyrimidinyloxy)aniline

Hazard: Irritant

Formula: C₁₀H₇ClFN₃O
Melting Pt: 134…136 °C
MDL Number: MFCD08056616
CAS Number: 213675-94-6

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR