2-Bromo-1-(4-chloro-2-fluoro-5-methylphenyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-fluoro-5-methylphenacyl bromide

043869-500MG 043869-1G 043869-5G
101862-986EA 278.46 USD
101862-986 101862-988 101862-990
2-Bromo-1-(4-chloro-2-fluoro-5-methylphenyl)-1-ethanone ≥95%
2-Bromo-1-(4-chloro-2-fluoro-5-methylphenyl)-1-ethanone

Hazard: Irritant

Formula: C₉H₇BrClFO
MW: 265.52 g/mol
Melting Pt: 78…81 °C
MDL Number: MFCD01935937
CAS Number: 338982-26-6

Order Now

Specification Test Results

Melting point 78-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR