1-Methyl-1H-indol-2-ol ≥95%

Supplier: Matrix Scientific

043867-500MG 043867-1G 043867-5G
101862-980EA 323.08 USD
101862-980 101862-982 101862-984
1-Methyl-1H-indol-2-ol ≥95%
1-Methyl-1H-indol-2-ol

Hazard: Irritant

Formula: C₉H₉NO
Melting Pt: 88…91 °C
MDL Number: MFCD01935997
CAS Number: 90563-58-9

Order Now

Specification Test Results

Melting point 88-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR