3-Phenyl-1H-indole-2-carbohydrazide ≥95%

Supplier: Matrix Scientific

043866-500MG 043866-1G
101862-976EA 1109.23 USD
101862-976 101862-978
3-Phenyl-1H-indole-2-carbohydrazide ≥95%
3-Phenyl-1H-indole-2-carbohydrazide

Hazard: Irritant

Formula: C₁₅H₁₃N₃O
Melting Pt: 238…240 °C
MDL Number: MFCD00974217
CAS Number: 105492-12-4

Order Now

Specification Test Results

Melting point 238-240°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR