Ethyl-2-(2-benzoylanilino)-2-oxoacetate ≥95%
Supplier: Matrix Scientific
043863-500MG
043863-1G
043863-5G
101862-964EA
375.38
USD
101862-964
101862-966
101862-968
Ethyl-2-(2-benzoylanilino)-2-oxoacetate ≥95%
Ethyl-2-(2-benzoylanilino)-2-oxoacetate
Hazard: Irritant
Formula:
C₁₇H₁₅NO₄ Melting Pt: 122…124 °C |
MDL Number:
MFCD00974203 CAS Number: 338401-00-6 |
Specification Test Results
Melting point | 122-124°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...