2-Amino-4-(2-naphthyl)-1H-pyrrole-3-carbonitrile ≥95%

Supplier: Matrix Scientific

043861-500MG 043861-1G
101862-956EA 1483.08 USD
101862-956 101862-958
2-Amino-4-(2-naphthyl)-1H-pyrrole-3-carbonitrile ≥95%
2-Amino-4-(2-naphthyl)-1H-pyrrole-3-carbonitrile

Hazard: Irritant

Formula: C₁₅H₁₁N₃
Melting Pt: 220…222 °C
MDL Number: MFCD00974200
CAS Number: 338400-97-8

Order Now

Specification Test Results

Melting point 220-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR