5-Nitro-2-pyridinethiol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Mercapto-5-nitropyridine, 5-Nitro-2-pyridyl Mercaptan, 5-Nitro-2-pyridinethiol

043857-500MG 043857-1G 043857-5G
101862-934EA 16.92 USD
101862-934 101862-936 101862-938
5-Nitro-2-pyridinethiol ≥95%
5-Nitro-2-pyridinethiol

Hazard: Irritant

Formula: C₅H₄N₂O₂S
MW: 156.16 g/mol
Melting Pt: 169…173 °C
MDL Number: MFCD00030890
CAS Number: 2127-09-5

Order Now

Specification Test Results

Melting point 169-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR