Ethyl-2-[(2-phenylacetyl)amino]acetate ≥95%
Supplier: Matrix Scientific
043856-500MG
043856-1G
043856-5G
101862-928EA
181.54
USD
101862-928
101862-930
101862-932
Ethyl-2-[(2-phenylacetyl)amino]acetate ≥95%
Ethyl-2-[(2-phenylacetyl)amino]acetate
Hazard: Irritant
Formula:
C₁₂H₁₅NO₃ Melting Pt: 83…85 °C |
MDL Number:
MFCD00793785 CAS Number: 4838-35-1 |
Specification Test Results
Melting point | 83-85°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...