Dimethyl-2-(4-chlorophenoxy)malonate ≥95%
Supplier: Matrix Scientific
043855-500MG
043855-1G
043855-5G
101862-922EA
181.54
USD
101862-922
101862-924
101862-926
Dimethyl-2-(4-chlorophenoxy)malonate ≥95%
Dimethyl-2-(4-chlorophenoxy)malonate
Hazard: Irritant
Formula:
C₁₁H₁₁ClO₅ Melting Pt: 77…79 °C |
MDL Number:
MFCD00793776 CAS Number: 338400-09-2 |
Specification Test Results
Melting point | 77-79°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...